Acenaphthene, 99%, Thermo Scientific Chemicals
Acenaphthene, 99%, Thermo Scientific Chemicals
Acenaphthene, 99%, Thermo Scientific Chemicals
Acenaphthene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acenaphthene, 99%, Thermo Scientific Chemicals
CAS: 83-32-9 | C12H10 | 154.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
View all
Plastic bottle
Catalog number ACR201341000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcenaphthene
CAS83-32-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0