Acenaphthene, 99%, Thermo Scientific Chemicals
Acenaphthene, 99%, Thermo Scientific Chemicals
Acenaphthene, 99%, Thermo Scientific Chemicals
Acenaphthene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acenaphthene, 99%, Thermo Scientific Chemicals
CAS: 83-32-9 | C12H10 | 154.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
View all
Glass Bottle
Catalog number ACR201340050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1427-06-1
IUPAC Namemethyl 6-fluoropyridine-3-carboxylate
Molecular FormulaC7H6FNO2
InChI KeyHLYBWNNPVXFCPZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CN=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%