Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
CAS: 13598-36-2 | H3O3P
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2 kg
Packaging:
Glass bottle
Catalog number ACR201155000
Price (MYR)
663.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
663.00
EA
Add to cart
Chemical Identifiers
CAS131674-41-4
IUPAC Name3-(2-methoxypyridin-3-yl)prop-2-enoic acid
Molecular FormulaC9H9NO3
InChI KeyMTVRDAQJMCORAQ-UHFFFAOYSA-N
SMILESCOC1=C(C=CC(O)=O)C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (unspecified)>94.0%
Appearance (Color)Grey