Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
CAS: 13598-36-2 | H3O3P
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
1 kg
Packaging:
Glass bottle
Catalog number ACR201150020
Price (MYR)
1,174.00
Add to cart
Quantity:
2 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,174.00
Add to cart
Specifications
Chemical Name or MaterialPhosphorous acid
ColorWhite
CAS13598-36-2
Melting Point73.0°C
Boiling Point200.0°C