Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals
CAS: 13598-36-2 | H3O3P
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
2 kg
Packaging:
Glass bottle
Catalog number ACR201150010
Price (MYR)
876.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
876.00
Add to cart
Chemical Identifiers
CAS20765-98-4
IUPAC Namerhodium(3+) trichloride
Molecular FormulaCl3Rh
InChI KeySONJTKJMTWTJCT-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Rh+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.