Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR201145000
Price (MYR)
543.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
543.00
Add to cart
Chemical Identifiers
CAS16940-66-2
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<5 ppm
Silicate (as SiO2)=<50 ppm
Fines=<1.0 %
Appearance (Color)White
Magnesium carbonate=<100 ppm