Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, for analysis, 85 wt% solution in water, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
Glass bottle
Catalog number ACR201140025
Price (MYR)
1,092.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,092.00
Add to cart
Specifications
Chemical Name or MaterialPhosphoric acid
Name Note85 wt% Solution in Water
CAS7664-38-2
Melting Point21.0°C
Boiling Point158.0°C