Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
CAS: 1314-56-3 | O5P2 | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number ACR200892500
Price (MYR)
510.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
510.00
Add to cart
Chemical Identifiers
CAS1603-40-3
IUPAC Name3-methylpyridin-2-amine
Molecular FormulaC6H8N2
InChI KeyRGDQRXPEZUNWHX-UHFFFAOYSA-N
SMILESCC1=CC=CN=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Infrared spectrumConforms
Appearance (Form)Liquid after melting
Water=<0.8 %
Refractive index1.5813 to 1.5833 (20°C, 589 nm)
View more
Adsorbent
RUO – Research Use Only