Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
CAS: 1314-56-3 | O5P2 | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Glass bottle
Catalog number ACR200890025
Price (MYR)
1,312.00
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,312.00
Add to cart
Specifications
Chemical Name or MaterialPhosphorus pentoxide
Melting Point340.0°C to 360.0°C
CAS1314-56-3
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Adsorbent
RUO – Research Use Only