Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
CAS: 1314-56-3 | O5P2 | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
Packaging:
Glass bottle
Catalog number ACR200890010
Price (MYR)
873.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
873.00
Add to cart
Chemical Identifiers
CAS1345-25-1
IUPAC Nameoxoiron
Molecular FormulaFeO
InChI KeyUQSXHKLRYXJYBZ-UHFFFAOYSA-N
SMILESO=[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Adsorbent
RUO – Research Use Only