Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals
Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals
Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals
Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vanadyl(IV) acetylacetonate, 99%, Thermo Scientific Chemicals
CAS: 3153-26-2 | C10H14O5V | 265.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number ACR199590500
Price (MYR)
730.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
730.00
Add to cart
Chemical Identifiers
CAS57-71-6
IUPAC Name(3E)-3-(hydroxyimino)butan-2-one
Molecular FormulaC4H7NO2
InChI KeyFSEUPUDHEBLWJY-HWKANZROSA-N
SMILESCC(=O)C(\C)=N\O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Water Content (Karl Fischer Titration)≤0.75%