Selenious acid, 98%, Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
CAS: 7783-00-8 | H2O3Se | 128.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number ACR198872500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSelenious acid
ColorPink or White
CAS7783-00-8
Melting Point70.0°C
Health Hazard 1GHS Signal Word: Danger