Selenious acid, 98%, Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Selenious acid, 98%, Thermo Scientific Chemicals
CAS: 7783-00-8 | H2O3Se | 128.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number ACR198870500
Price (MYR)
565.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
565.00
Add to cart
Chemical Identifiers
CAS2774-84-7
IUPAC Nameundec-10-yn-1-ol
Molecular FormulaC11H20O
InChI KeyYUQZOUNRPZBQJK-UHFFFAOYSA-N
SMILESOCCCCCCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4535-1.4605 @ 20?C
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless to white to yellow
Assay (GC)≥95.0%