2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, Thermo Scientific Chemicals
CAS: 2033-24-1 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR198450250
Price (MYR)
514.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
514.00
Add to cart
Chemical Identifiers
CAS121-44-8
IUPAC Nametriethylamine
Molecular FormulaC6H15N
InChI KeyZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILESCCN(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Water=<0.05 %
Lead (Pb)=<0.02 ppm
UVat 254 nm A: =<0.125 (0.1 M in water)