Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Periodic acid, 99+%, extra pure, Thermo Scientific Chemicals
Periodic acid, 99+%, H5IO6, CAS Number-10450-60-9, pentahydroxidooxidoiodine, paraperiodic acid, orthoperiodic acid, acid, periodic, hexaoxoiodic 5-acid, tetrahydroxyiodous acid, pentahydroxy-l5-iodane oxide, periodic acid, periodic acid, cryst, unii-ak1d44l87g, 100g, 1.0% max. Iodic Acid
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number ACR198401000
Price (MYR)
983.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
983.00
Add to cart
Specifications
Chemical Name or MaterialPeriodic acid
ColorWhite to Yellow
CAS10450-60-9
Melting Point122.0°C
Health Hazard 1GHS Signal Word: Danger