Lead, +99%, powder, Thermo Scientific Chemicals
Lead, +99%, powder, Thermo Scientific Chemicals
Lead, +99%, powder, Thermo Scientific Chemicals
Lead, +99%, powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lead, +99%, powder, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR1981100101 kgHDPE Plastic bottle
Catalog number ACR198110010
Price (MYR)
680.00
Add to cart
Quantity:
1 kg
Packaging:
HDPE Plastic bottle
Request bulk or custom format
Price (MYR)
680.00
Add to cart
Chemical Identifiers
CAS20358-03-6
IUPAC Name5-bromo-1,3-benzothiazol-2-amine
Molecular FormulaC7H5BrN2S
InChI KeyZPUJTWBWSOOMRP-UHFFFAOYSA-N
SMILESNC1=NC2=CC(Br)=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%