4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
CAS: 92-66-0 | C12H9Br | 233.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR197981000
Price (MYR)
1,792.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,792.00
Add to cart
Chemical Identifiers
CAS1780-26-3
IUPAC Name4,6-dichloro-2-methylpyrimidine
Molecular FormulaC5H4Cl2N2
InChI KeyFIMUTBLUWQGTIJ-UHFFFAOYSA-N
SMILESCC1=NC(Cl)=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormSolid or powder
Assay (GC)96.0% min
ClarityClear colorless to pale yellow solution in chloroform, no particulates