S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals
S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals
S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals
S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
S-Methyl methanethiolsulfonate, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR197390050
Price (MYR)
706.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
706.00
Add to cart
Chemical Identifiers
CAS321746-49-0
IUPAC Nametriethylsulfanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Molecular FormulaC8H15F6NO4S3
InChI KeyBLODSRKENWXTLO-UHFFFAOYSA-N
SMILESCC[S+](CC)CC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay by NMR≥97.5%
Assay from Supplier's CofA≥97.5%
Water Content (Karl Fischer Titration)≤0.5%
Proton NMRConforms to structure