Molecular sieves 5A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 5A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 5A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 5A, 8 to 12 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 5A, 8 to 12 mesh, Thermo Scientific Chemicals
CAS: 69912-79-4 | AlNaO6Si2 | 202.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR197295000
Price (MYR)
679.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
679.00
Add to cart
Specifications
Bulk Density680 to 760g/L
Chemical Name or MaterialMolecular sieves 5A
ColorBeige to Gray or Brown
PackagingGlass bottle
Residual Water1.5% max. (550°C, 2 hrs.)
View more
Drying agent
RUO – Research Use Only