Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR197275000
Price (MYR)
864.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
864.00
Add to cart
Chemical Identifiers
CAS5794-13-8
IUPAC Name(2S)-2-amino-3-carbamoylpropanoic acid hydrate
Molecular FormulaC4H10N2O4
InChI KeyRBMGJIZCEWRQES-DKWTVANSSA-N
SMILESO.N[C@@H](CC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Other amino acids=<1 % (TLC)
Loss on drying11.5 % to 12.5 % (130°C, 3 hrs)
Arsenic (As)=<1 ppm
Appearance (Color)White
View more
Drying agent
RUO – Research Use Only