Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, 8 to 12 mesh, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
Packaging:
View all
Plastic drum
Catalog number ACR197270050
Price (MYR)
2,755.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
2,755.00
Add to cart
Chemical Identifiers
CAS25655-41-8
Molecular Formula(C6H9NO)nI2
SMILESII.*-CC(-*)N1CCCC1=O
MDL NumberMFCD00084483
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<8 % (5 g, 105°C)
Iodide (I)=<6.6 % (on dried substance)
Appearance (Form)Powder
Assay9.0 to 12.0 % (available iodine)
Heavy metals (as Pb)=<20 ppm
View more
Drying agent
RUO – Research Use Only