Molecular sieves 4A, 4 to 8 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 4 to 8 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 4 to 8 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 4 to 8 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, 4 to 8 mesh, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR197265000
Price (MYR)
646.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
646.00
Add to cart
Chemical Identifiers
CAS13280-61-0
IUPAC Name1-methyl-2-[(1E)-2-{4-[(1E)-2-(2-methylphenyl)ethenyl]phenyl}ethenyl]benzene
Molecular FormulaC24H22
InChI KeyQKLPIYTUUFFRLV-YTEMWHBBSA-N
SMILESCC1=CC=CC=C1C=CC1=CC=C(C=CC2=CC=CC=C2C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or yellow-green or pale green to green
FormCrystals or crystalline powder or powder
Assay (HPLC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)176-185°C
Drying agent
RUO – Research Use Only