Molecular sieves 4A, 4 to 8 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 4 to 8 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 4 to 8 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 4 to 8 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, 4 to 8 mesh, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
Packaging:
View all
Plastic drum
Catalog number ACR197260050
Price (MYR)
2,289.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
2,289.00
Add to cart
Chemical Identifiers
CAS632-69-9
IUPAC Namedisodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Molecular FormulaC20H2Cl4I4Na2O5
InChI KeyKCQREHTWEUECQT-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C1=C(I)C=C2C(OC3=C(I)C([O-])=C(I)C=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<10 % (105°C)
Appearance (Form)Crystalline powder
Absorption Ratio P-15/P+151.21 to 1.95
Appearance (Color)Bordeaux to red-brown
View more
Drying agent
RUO – Research Use Only