Cobalt(II) acetate tetrahydrate, 98-102%, ACS reagent
Cobalt(II) acetate tetrahydrate, 98-102%, ACS reagent
Cobalt(II) acetate tetrahydrate, 98-102%, ACS reagent
Cobalt(II) acetate tetrahydrate, 98-102%, ACS reagent
Thermo Scientific Chemicals
Cobalt(II) acetate tetrahydrate, 98-102%, ACS reagent
CAS: 6147-53-1 | C4H6CoO4·4H2O | 249.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR197101000100 gPlastic bottle
Catalog number ACR197101000
Price (MYR)
882.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
882.00
Add to cart
Chemical Identifiers
CAS104-85-8
IUPAC Name4-methylbenzonitrile
Molecular FormulaC8H7N
InChI KeyVCZNNAKNUVJVGX-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid or solid
Infrared spectrumConforms
GC>=98.0 %