Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals
Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals
Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals
Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals
CAS: 7681-11-0 | IK | 166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Catalog number ACR196731000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7440-43-9
IUPAC Namecadmium(2+) ion
Molecular FormulaCd
InChI KeyWLZRMCYVCSSEQC-UHFFFAOYSA-N
SMILES[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.05mm (0.002in) thick.
Appearance (Color)White silvery
Assay from Supplier's CofA≥99.999% (metals basis)