Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
CAS: 16872-11-0 | BF4H | 87.812 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR196705000
Price (MYR)
834.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
834.00
Add to cart
Chemical Identifiers
CAS132327-80-1
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-[(triphenylmethyl)carbamoyl]butanoic acid
Molecular FormulaC39H34N2O5
InChI KeyWDGICUODAOGOMO-UHFFFAOYNA-N
SMILESOC(=O)C(CCC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
C-X bond formation reagent
RUO – Research Use Only