Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fluoroboric acid, 50 wt.% solution in water, Thermo Scientific Chemicals
CAS: 16872-11-0 | BF4H | 87.812 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR196700250
Price (MYR)
525.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
525.00
Add to cart
Specifications
Chemical Name or MaterialFluoroboric acid
ColorWhite
CAS16872-11-0
Melting Point-90.0°C
Boiling Point130.0°C
View more
C-X bond formation reagent
RUO – Research Use Only