Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3
Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3
Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3
Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3
Thermo Scientific Chemicals
Sodium standard solution, for AAS, 1 mg/ml Na in 0.5N HNO3
CAS: 7631-99-4 | NNaO3 | 84.99 g/mol
Have Questions?
Catalog NumberQuantity
ACR196311000100 mL
Catalog number ACR196311000
Price (MYR)
387.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
387.00
Add to cart
Chemical Identifiers
CAS87199-14-2
IUPAC Name[2-(hydroxymethyl)phenyl]boronic acid
Molecular FormulaC7H9BO3
InChI KeyAFHOBSCDNXGFMO-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
for atomic adsorption spectroscopy
RUO – Research Use Only