Phosphorus standard solution, for AAS, 1 mg/ml P in water
Phosphorus standard solution, for AAS, 1 mg/ml P in water
Phosphorus standard solution, for AAS, 1 mg/ml P in water
Phosphorus standard solution, for AAS, 1 mg/ml P in water
Thermo Scientific Chemicals
Phosphorus standard solution, for AAS, 1 mg/ml P in water
CAS: 7723-14-0 | P | 30.97 g/mol
Have Questions?
Catalog NumberQuantity
ACR196185000500 mL
Catalog number ACR196185000
Price (MYR)
748.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
748.00
Add to cart
Chemical Identifiers
CAS128-09-6
IUPAC Name1-chloropyrrolidine-2,5-dione
Molecular FormulaC4H4ClNO2
InChI KeyJRNVZBWKYDBUCA-UHFFFAOYSA-N
SMILESClN1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(5 % in acetone) Clear to light hazy colorless to light yellow
For atomic adsorption spectroscopy
RUO – Research Use Only