Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF
Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF
Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF
Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF
Thermo Scientific Chemicals
Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF
CAS: 7440-58-6 | Hf | 178.49 g/mol
Have Questions?
Catalog NumberQuantity
ACR196011000100 mL
Catalog number ACR196011000
Price (MYR)
513.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
513.00
Add to cart
Chemical Identifiers
CAS1822-51-1
IUPAC Namehydrogen 4-(chloromethyl)pyridine chloride
Molecular FormulaC6H7Cl2N
InChI KeyZDHKVKPZQKYREU-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClCC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Yellow to orange
Titration with HClO4>=96.0 %
For atomic adsorption spectroscopy
RUO – Research Use Only