Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ACR195935000
Price (MYR)
723.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
723.00
Add to cart
Chemical Identifiers
CAS7787-58-8
IUPAC Namebismuth(3+) tribromide
Molecular FormulaBiBr3
InChI KeyTXKAQZRUJUNDHI-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)Light yellow to yellow or light grey
Assay96 to 103 % (Bi)
Melting point218°C to 220°C
Chloride (Cl)=<0.1 %
View more
For atomic adsorption spectroscopy
RUO – Research Use Only