Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO3, Thermo Scientific Chemicals
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO<sub>3</sub>, Thermo Scientific Chemicals
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO<sub>3</sub>, Thermo Scientific Chemicals
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO<sub>3</sub>, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO3, Thermo Scientific Chemicals
CAS: 10141-05-6 | CoN2O6 | 182.941 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ACR195921000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS104-03-0
IUPAC Name2-(4-nitrophenyl)acetic acid
Molecular FormulaC8H7NO4
InChI KeyYBADLXQNJCMBKR-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige
Melting point153°C to 156°C
Titration with NaOH>=98.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
View more
For atomic adsorption spectroscopy
RUO – Research Use Only