Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO3, Thermo Scientific Chemicals
Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO<sub>3</sub>, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO3, Thermo Scientific Chemicals
CAS: 7440-70-2 | Ca | 40.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ACR195881000
Price (MYR)
346.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
346.00
Add to cart
Chemical Identifiers
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<500 ppm
UVat 260 nm A: =<0.05
Infrared spectrumConforms
Appearance (Form)Crystalline powder or flakes
GC>=98.5 %
View more
for atomic adsorption spectroscopy
RUO – Research Use Only