Dibutyltin dichloride, 96%, Thermo Scientific Chemicals
Dibutyltin dichloride, 96%, Thermo Scientific Chemicals
Dibutyltin dichloride, 96%, Thermo Scientific Chemicals
Dibutyltin dichloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibutyltin dichloride, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Packaging:
Glass bottle
Catalog number ACR194872500
Price (MYR)
1,137.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,137.00
Add to cart
Chemical Identifiers
CAS79-21-0
SpecificationsSpecification SheetSpecification Sheet
Titration with Na2S2O334.0 to 39.0 % (PAA content)
Appearance (Color)Clear colorless to light yellow-green
Appearance (Form)Solution
Titration11.0 to 15.0 % (H2O2 content)