Dibutyltin dichloride, 96%, Thermo Scientific Chemicals
Dibutyltin dichloride, 96%, Thermo Scientific Chemicals
Dibutyltin dichloride, 96%, Thermo Scientific Chemicals
Dibutyltin dichloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibutyltin dichloride, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
Packaging:
Glass bottle
Catalog number ACR194870250
Price (MYR)
444.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
444.00
Add to cart
Chemical Identifiers
CAS1835-49-0
IUPAC Nametetrafluorobenzene-1,4-dicarbonitrile
Molecular FormulaC8F4N2
InChI KeyPCRSJGWFEMHHEW-UHFFFAOYSA-N
SMILESFC1=C(F)C(C#N)=C(F)C(F)=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow-green to pale brown
Assay (HPLC)>97.5%