Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
CAS: 546-68-9 | C12H28O4Ti | 284.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR194702500
Price (MYR)
560.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
560.00
Add to cart
Chemical Identifiers
CAS28425-04-9
IUPAC Namedichloropalladium; bis(triethylphosphane)
Molecular FormulaC12H30Cl2P2Pd
InChI KeyULYNIEUXPCUIEL-UHFFFAOYSA-L
SMILESCl[Pd]Cl.CCP(CC)CC.CCP(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Powder
Infrared spectrumConforms
Palladium (Pd)>=24.9 %
Lewis acid
RUO – Research Use Only