Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
CAS: 546-68-9 | C12H28O4Ti | 284.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR194700050
Price (MYR)
455.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
455.00
Add to cart
Chemical Identifiers
CAS10101-97-0
IUPAC Namenickel(2+) hexahydrate sulfate
Molecular FormulaH12NiO10S
InChI KeyRRIWRJBSCGCBID-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.03% max.
Lewis acid
RUO – Research Use Only