Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals
CAS: 546-68-9 | C12H28O4Ti | 284.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number ACR194700025
Price (MYR)
2,168.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,168.00
Add to cart
Chemical Identifiers
CAS1336-21-6
IUPAC Nameammonium hydroxide
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES[NH4+].[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
KMnO4-reducing substance=<0.001 % (as O)
Phosphate (PO4)=<0.0001 %
Silver (Ag)=<0.000002 %
Barium (Ba)=<0.00001 %
View more
Lewis acid
RUO – Research Use Only