Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Thermo Scientific Chemicals
Bismuth(III) oxide, 98+%, pure
CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol
Have Questions?
Catalog number ACR192620050
Price (MYR)
5,250.00
Add to cart
Request bulk or custom format
Price (MYR)
5,250.00
Add to cart
Chemical Identifiers
CAS80-18-2
IUPAC Namemethyl benzenesulfonate
Molecular FormulaC7H8O3S
InChI KeyCZXGXYBOQYQXQD-UHFFFAOYSA-N
SMILESCOS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Refractive index1.5150 to 1.5180 (20°C, 589 nm)
Appearance (Form)Liquid
HPLC>=98.5 %
Infrared spectrumConforms
For ultra trace analysis
RUO – Research Use Only