Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Bismuth(III) oxide, 98+%, pure
Thermo Scientific Chemicals
Bismuth(III) oxide, 98+%, pure
CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol
Have Questions?
Catalog NumberQuantity
ACR1926200101 kg
Catalog number ACR192620010
Price (MYR)
1,422.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,422.00
Add to cart
Chemical Identifiers
CAS2396-84-1
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or viscous liquid
Refractive Index1.4930-1.4970 @ 20°C
Appearance (Color)Clear white to yellow
Assay (GC)≥95.0%