Tartrazine, 89%, pure
Tartrazine, 89%, pure
Tartrazine, 89%, pure
Tartrazine, 89%, pure
Thermo Scientific Chemicals
Tartrazine, 89%, pure
CAS: 1934-21-0 | C16H9N4Na3O9S2 | 534.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR191895000
Price (MYR)
939.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
939.00
Add to cart
Chemical Identifiers
CAS555-21-5
IUPAC Name2-(4-nitrophenyl)acetonitrile
Molecular FormulaC8H6N2O2
InChI KeyPXNJGLAVKOXITN-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(CC#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)111.0-118.0?C