Tartrazine, 89%, pure
Tartrazine, 89%, pure
Tartrazine, 89%, pure
Tartrazine, 89%, pure
Thermo Scientific Chemicals
Tartrazine, 89%, pure
CAS: 1934-21-0 | C16H9N4Na3O9S2 | 534.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR191891000
Price (MYR)
536.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
536.00
Add to cart
Specifications
Chemical Name or MaterialTartrazine
CAS1934-21-0
Health Hazard 1GHS Signal Word: Danger
Decomposition Information>300°C
Melting Point>300°C