Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Thermo Scientific Chemicals
Fluorescein diacetate, 97%, pure
CAS: 596-09-8 | C24H16O7 | 416.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR191660250
Price (MYR)
1,476.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,476.00
Add to cart
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES[H+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Strength0.0998 to 0.1002 N (20°C)
Appearance (Form)Solution