Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Thermo Scientific Chemicals
Fluorescein diacetate, 97%, pure
CAS: 596-09-8 | C24H16O7 | 416.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR191660050
Price (MYR)
555.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
555.00
Add to cart
Chemical Identifiers
CAS885278-10-4
IUPAC Name2-(propan-2-yloxy)pyridine-3-carbaldehyde
Molecular FormulaC9H11NO2
InChI KeyGNTQOKGIVMJHQG-UHFFFAOYSA-N
SMILESCC(C)OC1=C(C=O)C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
FormLiquid
Assay (HPLC)>97.5%