2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR190560250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromo-6-methoxynaphthalene
Name Note98%
CAS5111-65-9
CAS Min %97.5
CAS Max %100.0