2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-6-methoxynaphthalene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR190560050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS529-28-2
IUPAC Name1-iodo-2-methoxybenzene
Molecular FormulaC7H7IO
InChI KeyDVQWNQBEUKXONL-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.6220 to 1.6240 (20°C, 589 nm)
Appearance (Color)Clear yellow