Mercurochrome, 24-27% Mercury, Thermo Scientific Chemicals
Mercurochrome, 24-27% Mercury, Thermo Scientific Chemicals
Mercurochrome, 24-27% Mercury, Thermo Scientific Chemicals
Mercurochrome, 24-27% Mercury, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercurochrome, 24-27% Mercury, Thermo Scientific Chemicals
CAS: 129-16-8 | C20H8Br2HgNa2O6 | 750.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR190110250
Price (MYR)
612.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
612.00
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.5% (metals basis)
FormFoil, 0.787mm (0.031in) thick, annealed.