Lissamine Green B, pure
Lissamine Green B, pure
Lissamine Green B, pure
Lissamine Green B, pure
Thermo Scientific Chemicals
Lissamine Green B, pure
CAS: 3087-16-9 | C27H25N2NaO7S2 | 576.614 g/mol
Have Questions?
Catalog NumberQuantity
ACR19010025025 g
Catalog number ACR190100250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLissamine Green B
CAS3087-16-9
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular FormulaC27H25N2NaO7S2