1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR190025000
Price (MYR)
1,224.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,224.00
Add to cart
Chemical Identifiers
CAS108-99-6
IUPAC Name3-methylpyridine
Molecular FormulaC6H7N
InChI KeyITQTTZVARXURQS-UHFFFAOYSA-N
SMILESCC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Water Content≤0.5%
Appearance (Color)Clear colorless to yellow
Assay from Supplier's CofA≥98.5%