1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR190021000
Price (MYR)
471.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
471.00
Add to cart
Chemical Identifiers
CAS108-75-8
IUPAC Name2,4,6-trimethylpyridine
Molecular FormulaC8H11N
InChI KeyBWZVCCNYKMEVEX-UHFFFAOYSA-N
SMILESCC1=CC(C)=NC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow
Assay (GC)≥98.5%
Identification (FTIR)Conforms
CommentMay darken on storage
Refractive Index1.4965-1.5005 @ 20?C